Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD22 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989314
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9314 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9314 Supplier Bioss Supplier No. BS9748R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANKRD22 is a protein coding gene.

Specifications

Antigen ANKRD22
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ANKRD22
Gene Accession No. Q5VYY1
Gene Alias 5430429D21Rik; ANKRD22; ankyrin repeat domain 22; Ankyrin repeat domain-containing protein 22; D19Ertd675e
Gene Symbols ANKRD22
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ANKRD22.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118932
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.