Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANKRD23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557570
Description
Immunogen sequence: KRLRHRVPPR KPEPLVKPQS QAQVEPVGLE MFLKAAAENQ EYLIDKYLTD GGDPNAHDKL HRTALHWAC Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.
This gene is a member of the muscle ankyrin repeat protein (MARP) family and encodes a protein with four tandem ankyrin-like repeats. The protein is localized to the nucleus, functioning as a transcriptional regulator. Expression of this protein is induced during recovery following starvation. [provided by RefSeq].
Specifications
ANKRD23 | |
Polyclonal | |
Unconjugated | |
ANKRD23 | |
1110058D09Rik; 2310041C05Rik; AI853437; Ankrd23; ankyrin repeat domain 23; ankyrin repeat domain-containing protein 23; DARP; diabetes related ankyrin repeat protein; diabetes-realted ankyrin repeat protein; diabetes-related ankyrin repeat protein; MARP3; Muscle ankyrin repeat protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
200539 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86SG2 | |
ANKRD23 | |
Recombinant protein corresponding to Human ANKRD23. Recombinant protein control fragment (Product #RP-96177). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction