Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANKRD23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113086
Description
ANKRD23 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene is a member of the muscle ankyrin repeat protein (MARP) family and encodes a protein with four tandem ankyrin-like repeats. The protein is localized to the nucleus, functioning as a transcriptional regulator. Expression of this protein is induced during recovery following starvation. [provided by RefSeq].
Specifications
ANKRD23 | |
Polyclonal | |
Unconjugated | |
ANKRD23 | |
1110058D09Rik; 2310041C05Rik; AI853437; Ankrd23; ankyrin repeat domain 23; ankyrin repeat domain-containing protein 23; DARP; diabetes related ankyrin repeat protein; diabetes-realted ankyrin repeat protein; diabetes-related ankyrin repeat protein; MARP3; Muscle ankyrin repeat protein 3 | |
Rabbit | |
Protein G | |
RUO | |
200539, 78321 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q812A3, Q86SG2 | |
ANKRD23 | |
Recombinant Human Ankyrin repeat domain-containing protein 23 protein (1-200AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction