Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANKRD23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA535853
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Mouse (86%), Rat (82%), Pig (85%).
This gene is a member of the muscle ankyrin repeat protein (MARP) family and encodes a protein with four tandem ankyrin-like repeats. The protein is localized to the nucleus, functioning as a transcriptional regulator. Expression of this protein is induced during recovery following starvation. [provided by RefSeq].
Specifications
| ANKRD23 | |
| Polyclonal | |
| Unconjugated | |
| ANKRD23 | |
| 1110058D09Rik; 2310041C05Rik; AI853437; Ankrd23; ankyrin repeat domain 23; ankyrin repeat domain-containing protein 23; DARP; diabetes related ankyrin repeat protein; diabetes-realted ankyrin repeat protein; diabetes-related ankyrin repeat protein; MARP3; Muscle ankyrin repeat protein 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 200539 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.6 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q86SG2 | |
| ANKRD23 | |
| Recombinant protein encompassing a sequence within the center region of human ANKRD23. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction