Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559240
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559240 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559240 Supplier Invitrogen™ Supplier No. PA559240
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DNAKLKVTVK KQMDKIEELQ KNLLNANLSE DEKEQLKKLM ELKQSLECNL DQEMKKNVEL EREITGFKNL Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 56%.

ANKRD26 encodes a protein containing N-terminal ankyrin repeats which function in protein-protein interactions. Mutations in this gene are associated with autosomal dominant thrombocytopenia-2. Pseudogenes of this gene are found on chromosome 7, 10, 13 and 16. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD26
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKRD26
Gene Accession No. Q9UPS8
Gene Alias 5730521P14Rik; ANKRD26; ankyrin repeat domain 26; ankyrin repeat domain-containing protein 26; bA145E8.1; BC008111; KIAA1074; mKIAA1074; RGD1310736; THC2
Gene Symbols ANKRD26
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANKRD26. Recombinant protein control fragment (Product #RP-97038).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22852
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.