Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKRD27 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP150029R
Description
ANKRD27 Polyclonal antibody specifically detects ANKRD27 in Human samples. It is validated for Western BlotSpecifications
ANKRD27 | |
Polyclonal | |
Western Blot | |
ankyrin repeat domain 27 (VPS9 domain), ankyrin repeat domain-containing protein 27, DKFZp434L0718, FLJ00040, VARP, Vps9 domain and ankyrin-repeat-containing protein, VPS9 domain-containing protein, VPS9-ankyrin-repeat protein | |
The immunogen for this product maps to a region between residue 225 and 275 of human Vps9 Domain and Ankyrin-Repeat-Containing Protein using the numbering given in entry NP_115515.2 (GeneID 84079). | |
0.1 mL | |
Signal Transduction | |
84079 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction