Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109392
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109392 Supplier Invitrogen™ Supplier No. PA5109392
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAFLKLRDQP SLVQAIFNGD PDEVRALIFK KEDVNFQDNE KRTPLHAAAY LGDAEIIELL ILSGARVNAK DSKWLTPLHR AVASCSEEAV QVLLKHSADV NARDKNWQTP LHIAAANKAV KCAEALVPLL SNVNVSDRAG RTALHHAAFS GHGEMVKLLL SRGANINAFD KKDRRAIHWA AYMGHIEVVK LLVSHGAEVT CKDKKSYTPL HAAASSGMIS VVKYLLDLGV DMNEPNAYGN TPLHVACYNG QDVVVNELID CGAIVNQKNE KGFTPLHFAA ASTHGALCLE LLVGNGADVN.

Putative regulatory subunit of protein phosphatase 6 (PP6) that may be involved in the recognition of phosphoprotein substrates. Involved in the PP6-mediated dephosphorylation of NFKBIE opposing its degradation in response to TNF-alpha. Selectively inhibits the phosphatase activity of PPP1C. Targets PPP1C to modulate HNRPK phosphorylation. ANKRD28 selectively regulated phosphorylation of hnRNP-K at ser284, an effect that was enhanced in vivo by an S1013A/S1017A double mutant, suggesting that ANKRD28 targets PP1 in a regulated manner to dephosphorylate hnRNP-K selectively at ser284.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD28
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ANKRD28
Gene Accession No. O15084, Q505D1
Gene Alias AI465466; AI604979; ANKRD28; ankyrin repeat domain 28; ankyrin repeat domain-containing protein 28; E430019N21Rik; KIAA0379; mKIAA0379; phosphatase interactor targeting K protein; phosphatase interactor targeting protein hnRNP K; PITK; PP6-ARS-A; PPP1R65; protein phosphatase 1, regulatory subunit 65; protein phosphatase 6 ankyrin repeat subunit A; RGD1559931; serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit A; serine/threonine-protein phosphatase 6 regulatory subunit ARS-A
Gene Symbols ANKRD28
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ANKRD28 (NP_056014.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105522, 23243
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.