Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKRD30B Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305957B
Description
ANKRD30B Polyclonal specifically detects ANKRD30B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ANKRD30B | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Ankyrin Repeat Domain-Containing Protein 30B, Breast Cancer Antigen NY-BR-1.1, NY-BR-1.1, Serologically Defined Breast Cancer Antigen NY-BR-1.1 | |
Produced in rabbits immunized with E. coli-derived Human ANKRD30B fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Biotin | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
374860 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction