Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKRD32 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | ANKRD32 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ANKRD32 Polyclonal specifically detects ANKRD32 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ANKRD32 | |
Polyclonal | |
Rabbit | |
PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
ANKRD32 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Human | |
84250.0 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: SWLQMFVAEAVFKKLCLQSSGSVSSEPLSLQKMVYSYLPALGKTGVLGSGKIQVSKKIGQRPCFDSQRTLLMLNGTKQKQVEGLP | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title