Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559301
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559301 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559301 Supplier Invitrogen™ Supplier No. PA559301
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EYAKESKAMA IKTDVDVVEI KNTLRKHIYR AQAVRYNCIR IDKQPVYNVE VKNAEFPRGV LNLIESLIEG HFFKEAIEEL STLQAHYIPP VC Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

Plays a role in the DNA damage response (DDR) pathway by regulating postreplication repair of UV-damaged DNA and genomic stability maintenance (PubMed:25931565). The SLF1-SLF2 complex acts to link RAD18 with the SMC5-SMC6 complex at replication-coupled interstrand cross-links (ICL) and DNA double-strand breaks (DSBs) sites on chromatin during DNA repair in response to stalled replication forks (PubMed:25931565). Promotes the recruitment of SLF2 and the SMC5-SMC6 complex to DNA lesions (PubMed:25931565).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD32
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLF1
Gene Accession No. Q9BQI6
Gene Alias 2700017A04Rik; ANKRD32; ankyrin repeat domain 32; ankyrin repeat domain-containing protein 32; AW545819; BRCT domain containing 1; BRCT domain-containing protein 1; BRCTD1; Brctx; C730024G01Rik; Protein BRCTx; SLF1; Smc5/6 localization factor 1; SMC5-SMC6 complex localization factor 1; SMC5-SMC6 complex localization factor protein 1
Gene Symbols SLF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANKRD32. Recombinant protein control fragment (Product #RP-96854).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84250
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.