Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ ANKRD32 Polyclonal Antibody
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565851
Description
Immunogen sequence: SWLQMFVAEAV FKKLCLQSSG SVSSEPLSLQ KMVYSYLPAL GKTGVLGSGK IQVSKKIGQR PCFDSQRTLL MLNGTKQKQV EGLP Highest antigen sequence identity to the following orthologs - mouse 67%, rat 67%.
Plays a role in the DNA damage response (DDR) pathway by regulating postreplication repair of UV-damaged DNA and genomic stability maintenance (PubMed:25931565). The SLF1-SLF2 complex acts to link RAD18 with the SMC5-SMC6 complex at replication-coupled interstrand cross-links (ICL) and DNA double-strand breaks (DSBs) sites on chromatin during DNA repair in response to stalled replication forks (PubMed:25931565). Promotes the recruitment of SLF2 and the SMC5-SMC6 complex to DNA lesions (PubMed:25931565).
            Specifications
| ANKRD32 | |
| Polyclonal | |
| Unconjugated | |
| SLF1 | |
| 2700017A04Rik; ANKRD32; ankyrin repeat domain 32; ankyrin repeat domain-containing protein 32; AW545819; BRCT domain containing 1; BRCT domain-containing protein 1; BRCTD1; Brctx; C730024G01Rik; Protein BRCTx; SLF1; Smc5/6 localization factor 1; SMC5-SMC6 complex localization factor 1; SMC5-SMC6 complex localization factor protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84250 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | 
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BQI6 | |
| SLF1 | |
| Recombinant Human ANKRD32. Recombinant protein control fragment (Product #RP-106517). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
Safety and Handling
                                                    
                                                    WARNING: Cancer - www.P65Warnings.ca.gov 
                                                
                                            
                                        
                                    
                                
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction