Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD33 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559028
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559028 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559028 Supplier Invitrogen™ Supplier No. PA559028
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SKSVPELLGT APPPPLVPQS PPGSPQRSPW VFVPYQSPQG ILSKCLQWLQ PRDSTSPRPQ VPKILLSKAS SSSHQCQPKP SPSGHQSLAL PLW Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 60%.

Ankyrin repeat domain 33 (ANKRD33), also named as C12orf7, belongs to the ankyrin repeat family and is highly expressed in retina. ANKRD33 exists as two isoforms. The isoform 1 molecular weight is 29 kDa and isoform 2 is 49 kDa. The function of ANKRD33 is still non-known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD33
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKRD33
Gene Accession No. Q7Z3H0
Gene Alias A930021G21Rik; Ankrd33; ankyrin repeat domain 33; ankyrin repeat domain-containing protein 33; C12orf7; LOC683410; PANKY; Panky-a; Photoreceptor ankyrin repeat protein; photorecptor ankyrin repeat protein A; similar to ankyrin repeat domain 33
Gene Symbols ANKRD33
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANKRD33. Recombinant protein control fragment (Product #RP-97806).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 341405
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.