Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD34B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560326
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560326 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560326 Supplier Invitrogen™ Supplier No. PA560326
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HYSSDSQLSA GLTPPTSEDG KALIGKKKIL SPSPSQLSES KELLENIPPG PLSRRNHAVL ERRGSGAFPL DHSVTQTR Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.

ANKRD34B, also named as Ankyrin repeat domain-containing protein 34B, is a 514 amino acid protein, which contains 4 ANK repeats and belongs to the ANKRD34 family. The calculated molecular weight of ANKRD34B is 55 kDa, but the phosphorylated protein is about 60-65 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD34B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKRD34B
Gene Accession No. A5PLL1
Gene Alias 6430502M16Rik; ANKRD34B; ankyrin repeat domain 34B; ankyrin repeat domain-containing protein 34B; cytosolic phosphoprotein DP58; dendritic phosphoprotein 58 kDa; dendritic protein of 58 kDa; Dp58
Gene Symbols ANKRD34B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANKRD34B. Recombinant protein control fragment (Product #RP-100288).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340120
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.