Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANKRD36 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571030
Description
This target displays homology in the following species: Cow: 77%; Dog: 77%; Horse: 77%; Human: 100%.
ANKRD36 (Ankyrin Repeat Domain 36) is a Protein Coding gene. An important paralog of this gene isANKRD36C.
Specifications
ANKRD36 | |
Polyclonal | |
Unconjugated | |
ANKRD36 | |
ANKRD36; ankyrin repeat domain 36; ankyrin repeat domain-containing protein 36; ankyrin repeat domain-containing protein 36A; ankyrin-related; UNQ2430 | |
Rabbit | |
Affinity chromatography | |
RUO | |
375248 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A6QL64 | |
ANKRD36 | |
synthetic peptide directed towards the C-terminal region of human ANKRD36. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction