Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD36C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565680
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565680 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565680 Supplier Invitrogen™ Supplier No. PA565680
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KDEQISGTVSS QKQPALKATS DKKDSVSNIP TEIKDGQQSG TVSSQKQLAW KATSVKKDSV SNIATEIKDG QIRGTVS Highest antigen sequence identity to the following orthologs - mouse 28%, rat 28%.

ANKRD36C is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD36C
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKRD36C
Gene Accession No. Q5JPF3
Gene Alias ANKRD36C; ankyrin repeat domain 36C; ankyrin repeat domain-containing protein 36C; protein immuno-reactive with anti-PTH polyclonal antibodies
Gene Symbols ANKRD36C
Host Species Rabbit
Immunogen Recombinant Human ANKRD36C. Recombinant protein control fragment (Product #RP-104932).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 400986
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.