Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANKRD37 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570158
Description
This target displays homology in the following species: Cow: 93%; Dog: 86%; Goat: 93%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
ANKRD37 (Ankyrin Repeat Domain 37) is a Protein Coding gene. An important paralog of this gene isANKRD42.
Specifications
| ANKRD37 | |
| Polyclonal | |
| Unconjugated | |
| ANKRD37 | |
| Ankrd37; ankyrin repeat domain 37; ankyrin repeat domain-containing protein 37; hLrp2bp; low density lipoprotein receptor-related protein binding protein; low-density lipoprotein receptor-related protein 2-binding protein; LPR2BP; Lrp2bp; RGD1565715 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 654824 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q569N2 | |
| ANKRD37 | |
| synthetic peptide directed towards the middle region of human ANKRD37 (aa 46-95). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction