Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD39 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531671
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531671 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531671 Supplier Invitrogen™ Supplier No. PA531671
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32. Predicted reactivity: Human (99%), Mouse (92%), Rat (91%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ANKRD39, or Ankyrin Repeat Domain 39, is part of the ankyrin repeat domain (ANKRD) protein family. Ankyrin repeat domains are known for mediating protein-protein interactions and are involved in various cellular processes, including regulation of cell cycle, transcription, and signal transduction. The specific function of ANKRD39 is not yet fully elucidated, but its family members are typically regulators of other proteins' activity and stability. ANKRD proteins can bind to diverse sets of targets, contributing to their role in various physiological and pathological contexts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD39
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene ANKRD39
Gene Accession No. Q53RE8
Gene Alias 9130416N05Rik; ANKRD39; ankyrin repeat domain 39; ankyrin repeat domain-containing protein 39; C030004B10Rik; HSPC200
Gene Symbols ANKRD39
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ANKRD39. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51239
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.