Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD40 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989040
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9040 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9040 Supplier Bioss Supplier No. BS9399R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen ANKRD40
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ANKRD40
Gene Accession No. Q6AI12
Gene Alias ANKRD40; ankyrin repeat domain 40; Ankyrin repeat domain-containing protein 40
Gene Symbols ANKRD40
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ANKRD40.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91369
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.