Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD45 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556811
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556811 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556811 Supplier Invitrogen™ Supplier No. PA556811
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEDKNTILSA CRAKNEWLET HTEASINELF EQRQQLEDIV TPIFTKMTTP CQVKSAKSVT SHDQKRSQDD TSN Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 52%.

ANKRD45, or Ankyrin Repeat Domain 45, belongs to the large family of ankyrin repeat domain-containing proteins. Proteins with ankyrin repeats typically serve as scaffolds for protein-protein interactions, thus playing a critical role in various cellular processes, such as signal transduction, cell cycle regulation, and the maintenance of cellular structure. While the specific functions of ANKRD45 remain largely unexplored, studies on related ankyrin repeat proteins suggest that they may have roles in cellular stress responses and cancer development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD45
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKRD45
Gene Accession No. Q5TZF3
Gene Alias 4933409K03Rik; ANKRD45; ankyrin repeat domain 45; ankyrin repeat domain-containing protein 45; cancer/testis antigen 117; CT117; RGD1311153; RP3-436N22.4
Gene Symbols ANKRD45
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANKRD45. Recombinant protein control fragment (Product #RP-94499).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339416
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.