Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD46 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569962
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569962 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569962 Supplier Invitrogen™ Supplier No. PA569962
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 93%; Rat: 100%; Zebrafish: 79%.

ANKRD46 encodes a protein containing multiple ankyrin repeats. Ankyrin domains function in protein-protein interactions in a variety of cellular processes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD46
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ANKRD46
Gene Accession No. Q8BTZ5
Gene Alias 1110054N06Rik; AI314978; AI987733; ankrd46; ankrd46b; ANK-S; ankyrin repeat domain 46; ankyrin repeat domain 46b; ankyrin repeat domain-containing protein 46; Ankyrin repeat small protein; GENX-115279; hypothetical protein LOC157567-like protein; loc157567 protein; RGD1309382; wu:fq41d11; zgc:109722
Gene Symbols ANKRD46
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of mouse ANKRD46.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68839
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.