Learn More
Description
Specifications
Specifications
| Antigen | ANKRD5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000 |
| Formulation | PBS and 2% Sucrose with 0.09% Sodium Azide |
| Gene Alias | ankyrin repeat domain 5, ankyrin repeat domain-containing protein 5, dJ839B4.6, FLJ21669 |
| Gene Symbols | ANKRD5 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptides corresponding to ANKRD5 (ankyrin repeat domain 5) The peptide sequence was selected from the N terminal of ANKRD5. Peptide sequence MTIVDNEGKGVLFYCILPTKRHYRCALIALEHGADVNNSTYEGKPIFLRA. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
