Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKRD50 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP150032UV
Description
ANKRD50 Polyclonal antibody specifically detects ANKRD50 in Human, Mouse samples. It is validated for Western BlotSpecifications
ANKRD50 | |
Polyclonal | |
Western Blot | |
ankyrin repeat domain 50 | |
The immunogen for this product maps to a region between residue 1379 and 1429 of human Ankyrin Repeat Domain 50 using the numbering given in entry NP_065070.1 (GeneID 57182). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
57182 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction