Learn More
ANKRD50 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303000AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Involved in the endosome-to-plasma membrane trafficking and recycling of SNX27-retromer-dependent cargo proteins, such as GLUT1.Specifications
ANKRD50 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9ULJ7 | |
ANKRD50 | |
Between 1379 and 1429 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ANKRD50 | |
KIAA1223 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57182, 99696 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.