Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD50 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989317
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9317 Supplier Bioss Supplier No. BS9751R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Involved in the endosome-to-plasma membrane trafficking and recycling of SNX27-retromer-dependent cargo proteins, such as GLUT1.

Specifications

Antigen ANKRD50
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ANKRD50
Gene Accession No. Q9ULJ7
Gene Alias AI662170; ANKRD50; ankrin repeat domain 50; ankyrin repeat domain 50; Ankyrin repeat domain-containing protein 50; BOS_16381; E430012K20Rik; KIAA1223; mKIAA1223; mKIAA3019; RGD1311665
Gene Symbols ANKRD50
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ANKRD50.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57182
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.