Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD53 Monoclonal Antibody (OTI1B8), TrueMAB™, OriGene
SDP

Catalog No. 502541436
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1436 30 μL
50-167-3636 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1436 Supplier OriGene Supplier No. TA502597S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ANKRD53 Monoclonal antibody specifically detects ANKRD53 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen ANKRD53
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI1B8
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ANKRD53
Gene Accession No. Q8N9V6
Gene Alias 4930564N15Rik; ANKRD53; ankyrin repeat domain 53; ankyrin repeat domain-containing protein 53
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-300 of human ANKRD53 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79998
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.