Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKRD53 Monoclonal Antibody (OTI2E4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502595S


Description
ANKRD53 Monoclonal antibody specifically detects ANKRD53 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
ANKRD53 | |
Monoclonal | |
0.63 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8N6T3 | |
Mouse | |
Affinity chromatography | |
RUO | |
55738 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI2E4 | |
Unconjugated | |
ARFGAP1 | |
4930564N15Rik; ANKRD53; ankyrin repeat domain 53; ankyrin repeat domain-containing protein 53 | |
Full length human recombinant protein of human ARFGAP1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction