Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD54 Antibody, Novus Biologicals™
SDP

Catalog No. NBP157052 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP157052 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP157052 Supplier Novus Biologicals Supplier No. NBP157052
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANKRD54 Polyclonal specifically detects ANKRD54 in Human samples. It is validated for Western Blot.

Specifications

Antigen ANKRD54
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q6NXT1
Gene Alias ankyrin repeat domain 54, LIARankyrin repeat domain-containing protein 54
Gene Symbols ANKRD54
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ANKRD54 (ankyrin repeat domain 54) The peptide sequence was selected from the middle region of ANKRD54 (NP_620152). Peptide sequence EVHALKRLRDSANANDVETVQQLLEDGADPCAADDKGRTALHFASCNGND.
Molecular Weight of Antigen 33 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 129138
Test Specificity Expected identity based on immunogen sequence: Canine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 92%.
Reconstitution Centrifuge vial prior to reconstitution. Add 50μL distilled water to a final antibody concentration of 1mg/mL.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.