Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKRD57 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10068222
Description
ANKRD57 Polyclonal specifically detects ANKRD57 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ANKRD57 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
ankyrin repeat domain 57, ankyrin repeat domain-containing protein 57, C2orf26, FLJ21870 | |
Rabbit | |
Affinity Purified | |
RUO | |
65124 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
NP_075392.2 | |
SOWAHC | |
The immunogen recognized by this antibody maps to a region between residue 475 and 525 of human ankyrin repeat domain 57 using the numbering given in entry NP_075392.2 (GeneID 65124). | |
0.1 mL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction