Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565155 Supplier Invitrogen™ Supplier No. PA565155
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSRLRNVKVQT ALLPMNEAAR SDQQAGPCVN RGTQTKKSGK SGPTRHRAQQ PAASSTCGQP PPATGSEQTG PHIRDTSQAL ELTQYFFEAV STQMEKWYER KIEE Highest antigen sequence identity to the following orthologs - mouse 65%, rat 63%.

Recruits CKI-epsilon to the beta-catenin degradation complex that consists of AXN1 or AXN2 and GSK3-beta and allows efficient phosphorylation of beta-catenin, thereby inhibiting beta-catenin/Tcf signals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKRD6
Gene Accession No. Q9Y2G4
Gene Alias ANKRD6; ankyrin repeat domain 6; ankyrin repeat domain-containing protein 6; diego homolog; diversin; Kiaa0957
Gene Symbols ANKRD6
Host Species Rabbit
Immunogen Recombinant Human ANKRD6. Recombinant protein control fragment (Product #RP-105904).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22881
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.