Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANKRD65 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564240
Description
Immunogen sequence: EEEQELRWME LDSEEALGTR TEGPSVVQGW GHLLQAVWRG PAGLVTQLLR QGH Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 51%.
The function remain unknown.
Specifications
ANKRD65 | |
Polyclonal | |
Unconjugated | |
ANKRD65 | |
ANKRD65; ankyrin repeat domain 65; ankyrin repeat domain-containing protein 65; hCG_20426; LOC441869 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
441869 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
E5RJM6 | |
ANKRD65 | |
Recombinant protein corresponding to Human ANKRD65. Recombinant protein control fragment (Product #RP-103609). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction