Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD65 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543104
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543104 Supplier Invitrogen™ Supplier No. PA543104
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MDSQRPEPRE EEEEEQELRW MELDSEEALG TRTEGPSVVQ GWGHLLQAVW Sequence homology: Dog: 92%; Human: 100%.

The function remain unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD65
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ANKRD65
Gene Accession No. E5RJM6
Gene Alias ANKRD65; ankyrin repeat domain 65; ankyrin repeat domain-containing protein 65; hCG_20426; LOC441869
Gene Symbols ANKRD65
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human hCG_20426 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441869
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.