Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKRD9 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988166
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8166 Supplier Bioss Supplier No. BS7964R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANKRD9 gene ontology annotations related to this gene include post-translational protein modification; protein localization to plasma membrane.

Specifications

Antigen ANKRD9
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ANKRD9
Gene Accession No. Q96BM1
Gene Alias 2500003O20Rik; ANKRD9; ankyrin repeat domain 9; Ankyrin repeat domain-containing protein 9; BOS_20505
Gene Symbols ANKRD9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ANKRD9.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122416
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.