Learn More
Invitrogen™ ANKS1 Monoclonal Antibody (6B11)

Description
Peptide Sequence: HVFSTVDVNL TYEIILTLGQ AFEVAYQLAL QAQKSRATGA SAAEMIETKS SKPVPKPRVG VRKSALEPPD MDQDAQSHAS VSWVVDPKPD SKRSLS.
Specifications
Specifications
| Antigen | ANKS1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 6B11 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ANKS1A |
| Gene Accession No. | Q92625 |
| Gene Alias | ANKS1; ANKS1A; ankyrin repeat and SAM domain containing 1; ankyrin repeat and SAM domain-containing protein 1A; ankyrin repeat and sterile alpha motif domain containing 1; ankyrin repeat and sterile alpha motif domain containing 1A; KIAA0229; ODIN |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.