Learn More
Invitrogen™ ANKS1 Polyclonal Antibody

Description
Immunogen sequence: VVQILLAAGT DVNIKDNHGL TALDTVRELP SQKSQQIAAL IEDHMTGKRS TKEVDKTPPP QPPLISSMDS ISQKSQGDVE KAVTELIIDF DANAEEEGP Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.
Specifications
Specifications
| Antigen | ANKS1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ANKS1A |
| Gene Accession No. | Q92625 |
| Gene Alias | ANKS1; ANKS1A; ankyrin repeat and SAM domain containing 1; ankyrin repeat and SAM domain-containing protein 1A; ankyrin repeat and sterile alpha motif domain containing 1; ankyrin repeat and sterile alpha motif domain containing 1A; KIAA0229; ODIN |
| Gene Symbols | ANKS1A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.