Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANKS1A Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171806PE
Description
ANKS1A Polyclonal antibody specifically detects ANKS1A in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
ANKS1A | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
ANKS1ankyrin repeat and SAM domain-containing protein 1A, ankyrin repeat and SAM domain containing 1, ankyrin repeat and sterile alpha motif domain containing 1, ankyrin repeat and sterile alpha motif domain containing 1A, KIAA0229MGC42354, ODIN | |
The immunogen for this product maps to a region between residue 875 and 925 of human Ankyrin Repeat and Sterile Apha Motif Domain Containing 1A using the numbering given in entry NP_056060.2 (GeneID 23294). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
PE | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23294 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction