Learn More
ANKS1A Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303050A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
Regulator of different signaling pathways. Regulates EPHA8 receptor tyrosine kinase signaling to control cell migration and neurite retraction (By similarity).Specifications
ANKS1A | |
Polyclonal | |
Unconjugated | |
ANKS1A | |
ANKS1, ankyrin repeat and SAM domain containing 1, ankyrin repeat and sterile alpha motif domain containing 1, KIAA0229, Odin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23294 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q92625 | |
ANKS1A | |
Between 1084 and 1134 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.