Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKS1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545780
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545780 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545780 Supplier Invitrogen™ Supplier No. PA545780
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ACAKMRANCQ KSTEQMKKVP TIILSVSYKG VKFIDATNKN IIAEHEIRNI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

This gene encodes a multi-domain protein that is predominantly expressed in brain and testis. This protein interacts with amyloid beta protein precursor (AbetaPP) and may have a role in normal brain development, and in the pathogenesis of Alzheimer's disease. Expression of this gene has been shown to be elevated in patients with pre-B cell acute lymphocytic leukemia associated with t(1;19) translocation. Alternatively spliced transcript variants encoding different isoforms (some with different subcellular localization) have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKS1B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ANKS1B
Gene Accession No. Q7Z6G8
Gene Alias AIDA; AIDA-1; AIDA-1b; amyloid-beta precursor protein intracellular domain associated protein 1; amyloid-beta protein intracellular domain-associated protein 1; ANKS1B; ANKS2; ankyrin repeat and sterile alpha motif domain containing 1B; ankyrin repeat and sterile alpha motif domain-containing protein 1B; C030032C09Rik; cajalin 2; cajalin-2; E2A-PBX1-associated protein; E530015N03Rik; EB1; EB-1; ENSMUSG00000078431; Gm10937; Gm1555; MGC26087; RGD1565556
Gene Symbols ANKS1B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ANKS1B (aa 1084-1133).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56899
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.