Learn More
Invitrogen™ ANKS1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121141
Description
Positive test controls include: A-549, Mouse liver. The target is usually found in the following locations: Cajal body, Cell junction, Cell projection, Cytoplasm, Nucleus, dendritic spine, postsynaptic cell membrane, postsynaptic density, synapse. Immunogen sequence: GFPFSAIHQV HNTGDWGEPS ITLRPPNEAT ASTPVQYWQH HPEKLIFQSC DYKAFYLGSM LIKELRGTES TQDACAKMRA NCQKSTEQMK KVPTIILSVS YKGVKFIDAT NKNIIAEHEI RNISCAAQDP EDLSTFAYIT KDLKSNHHYC HVFTAFDVNL AYEIILTLGQ AFEVAYQLAL QARKGGHSST LPESFENKPS KPIPKPRVSI RKSVQIDPSE QKTLANLPWI VEPGQEAKRG INTKYETTIF.
This gene encodes a multi-domain protein that is predominantly expressed in brain and testis. This protein interacts with amyloid beta protein precursor (AbetaPP) and may have a role in normal brain development, and in the pathogenesis of Alzheimer's disease. Expression of this gene has been shown to be elevated in patients with pre-B cell acute lymphocytic leukemia associated with t(1;19) translocation. Alternatively spliced transcript variants encoding different isoforms (some with different subcellular localization) have been described for this gene.
Specifications
| ANKS1B | |
| Polyclonal | |
| Unconjugated | |
| ANKS1B | |
| AIDA; AIDA-1; AIDA-1b; amyloid-beta precursor protein intracellular domain associated protein 1; amyloid-beta protein intracellular domain-associated protein 1; ANKS1B; ANKS2; ankyrin repeat and sterile alpha motif domain containing 1B; ankyrin repeat and sterile alpha motif domain-containing protein 1B; C030032C09Rik; cajalin 2; cajalin-2; E2A-PBX1-associated protein; E530015N03Rik; EB1; EB-1; ENSMUSG00000078431; Gm10937; Gm1555; MGC26087; RGD1565556 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 314721, 56899, 77531 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P0C6S7, Q7Z6G8, Q8BIZ1 | |
| ANKS1B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 261-510 of human ANKS1B (NP_858056.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.