Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKS1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140298
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140298 Supplier Invitrogen™ Supplier No. PA5140298
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DSLGRTVLDI LKEHPSQKSL QIATLLQEYL EGVGRSTVLE EPVQEDATQE THISSPVESP SQKTKSETVT GELSKLLDEI KLCQEKDYSF EDLCHTISDH YL Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 89%.

This gene encodes a multi-domain protein that is predominantly expressed in brain and testis. This protein interacts with amyloid beta protein precursor (AbetaPP) and may have a role in normal brain development, and in the pathogenesis of Alzheimer's disease. Expression of this gene has been shown to be elevated in patients with pre-B cell acute lymphocytic leukemia associated with t(1;19) translocation. Alternatively spliced transcript variants encoding different isoforms (some with different subcellular localization) have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKS1B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKS1B
Gene Accession No. Q7Z6G8
Gene Alias AIDA; AIDA-1; AIDA-1b; amyloid-beta precursor protein intracellular domain associated protein 1; amyloid-beta protein intracellular domain-associated protein 1; ANKS1B; ANKS2; ankyrin repeat and sterile alpha motif domain containing 1B; ankyrin repeat and sterile alpha motif domain-containing protein 1B; C030032C09Rik; cajalin 2; cajalin-2; E2A-PBX1-associated protein; E530015N03Rik; EB1; EB-1; ENSMUSG00000078431; Gm10937; Gm1555; MGC26087; RGD1565556
Gene Symbols ANKS1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANKS1B. Recombinant protein control fragment (Product #RP-97868).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56899
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.