Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANKS3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559589
Description
Immunogen sequence: LNHGVKVDAR DHSGATARML AKQYGHMKIV ALMDTYSPSL PKSLYRSPEK YEDLSSSDES CPAPQRQRPC RKKGVSI Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.
May be involved in vasopressin signaling in the kidney. [UniProt].
Specifications
ANKS3 | |
Polyclonal | |
Unconjugated | |
ANKS3 | |
2700067D09Rik; Anks3; Ankyrin repeat and SAM domain-containing protein 3; ankyrin repeat and sterile alpha motif domain containing 3; C81345; KIAA1977; mKIAA1977; RGD1305833 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
124401 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZW76 | |
ANKS3 | |
Recombinant protein corresponding to Human ANKS3. Recombinant protein control fragment (Product #RP-98744). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction