Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKS4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560402
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560402 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560402 Supplier Invitrogen™ Supplier No. PA560402
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VALLDKAATA QNIMNPKKVT RLKEQAQKNA RRQIKECERL QEKHQNKMAH TYSKEESGTL SSSKGTFSRS SPSNASAP Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 82%.

As part of the intermicrovillar adhesion complex/IMAC plays a role in epithelial brush border differentiation, controlling microvilli organization and length. Plays a role in assembly of the complex (PubMed:26812018). May play a role in cellular response to endoplasmic reticulum stress. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKS4B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKS4B
Gene Accession No. Q8N8V4
Gene Alias ANKS4B; Ankyrin repeat and SAM domain-containing protein 4B; ankyrin repeat and sterile alpha motif domain containing 4B; harmonin-interacting ankyrin repeat-containing protein; harmonin-interacting ankyrin-repeat containing protein; HARP
Gene Symbols ANKS4B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANKS4B. Recombinant protein control fragment (Product #RP-98775).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257629
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.