Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKS6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552550
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552550 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552550 Supplier Invitrogen™ Supplier No. PA552550
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSTTSKSTSP TLTPSPSPKG HTAESSVSSS SSHRQSKSSG GSSSGTITDE DELTGILKKL SLEKYQPIFE EQEVDMEAFL TLTDGDLKEL GIKTDGSRQQ ILAAISELNA GKGRERQILQ ETIHNFHSSF Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

ANKS6 is required for renal function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKS6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANKS6
Gene Accession No. Q68DC2
Gene Alias 2210417J20Rik; Ankrd14; ANKS6; Ankyrin repeat and SAM domain-containing protein 6; ankyrin repeat and sterile alpha motif domain containing 6; ankyrin repeat domain 14; Ankyrin repeat domain-containing protein 14; b2b1801.1Clo; Gm635; NPHP16; PKDR1; polycystic kidney disease protein 1; SAM domain-containing protein 6; samcystin; SAMD6; sterile alpha motif domain containing 6; sterile alpha motif domain-containing protein 6
Gene Symbols ANKS6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANKS6. Recombinant protein control fragment (Product #RP-89528).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203286
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.