Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKS6 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988820
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8820 Supplier Bioss Supplier No. BS9099R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANKS6 is required for renal function (By similarity).

Specifications

Antigen ANKS6
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ANKS6
Gene Accession No. Q68DC2
Gene Alias ANKRD14; ANKS6; Ankyrin repeat and SAM domain-containing protein 6; ankyrin repeat and sterile alpha motif domain containing 6; ankyrin repeat domain 14; Ankyrin repeat domain-containing protein 14; NPHP16; PKDR1; SAM domain-containing protein 6; SamCystin; SAMD6; Sterile alpha motif domain-containing protein 6
Gene Symbols ANKS6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ANKS6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203286
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.