Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ankyrin 1 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171805FR
Description
Ankyrin 1 Polyclonal antibody specifically detects Ankyrin 1 in Human samples. It is validated for Western BlotSpecifications
Ankyrin 1 | |
Polyclonal | |
Western Blot | |
ANK, ANK-1, ankyrin 1, erythrocytic, ankyrin-1, Ankyrin-R, Erythrocyte ankyrin, SPH1, SPH2 | |
The immunogen for this product maps to a region between residue 1831 and 1881 of human Ankyrin 1 using the numbering given in entry NP_065209.2 (GeneID 286). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
286 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction