Learn More
Ankyrin 1 Antibody (S388A-60), DyLight 488, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259316G
Catalog No. NBP259316G
Only null left
Add to Cart
Description
Description
Ankyrin 1 Monoclonal specifically detects Ankyrin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Ankyrin 1
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
ANK1
Fusion protein amino acids 1-1881 (full-length) of human Ankyrin-R
100 μL
286
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
S388A-60
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
ANK, ANK-1, ankyrin 1, erythrocytic, ankyrin-1, Ankyrin-R, Erythrocyte ankyrin, SPH1, SPH2
Mouse
Protein G purified
Primary
Detects 200kDa. Cross-reacts with Ankyrin-B and Ankyrin-G.
Store at 4C in the dark.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.