Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ankyrin 1 Monoclonal Antibody (N388A/10), FITC
GREENER_CHOICE

Catalog No. PIMA545730
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545730 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545730 Supplier Invitrogen™ Supplier No. MA545730
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:100 dilution of MA5-45730 was sufficient for detection of Ankyrin R in 20 μg of mouse brain lysate by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 200kDa. Does not cross-react with Ankyrin-B or Ankyrin-G. This antibody was formerly sold as clone S388A-10.

Ankyrins are a family of proteins that link the integral membrane proteins to the underlying spectrin-actin cytoskeleton and play key roles in activities such as cell motility, activation, proliferation, contact and the maintenance of specialized membrane domains. Multiple isoforms of ankyrin with different affinities for various target proteins are expressed in a tissue-specific, developmentally regulated manner. Most ankyrins are typically composed of three structural domains: an amino-terminal domain containing multiple ankyrin repeats; a central region with a highly conserved spectrin binding domain; and a carboxy-terminal regulatory domain which is the least conserved and subject to variation. Ankyrin 1, the prototype of this family, was first discovered in the erythrocytes, but since has also been found in brain and muscles. Mutations in erythrocytic ankyrin 1 have been associated in approximately half of all patients with hereditary spherocytosis. Complex patterns of alternative splicing in the regulatory domain, giving rise to different isoforms of ankyrin 1 have been described. Truncated muscle-specific isoforms of ankyrin 1 resulting from usage of an alternate promoter have also been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ankyrin 1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N388A/10
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene ANK1
Gene Accession No. P16157, Q02357
Gene Alias ANK; ANK1; ANK-1; ANK-1; erythrocyte skeletal protein; ankyrin 1; ankyrin 1, erythrocytic; ankyrin 1, erythroid; ankyrin-1; Ankyrin-R; erythrocyte ankyrin; nb; normoblastic anemia; pale; rCG_43073; SPH1; SPH2
Gene Symbols ANK1
Host Species Mouse
Immunogen Fusion protein amino acids 1-1881 (full-length) of human Ankyrin-R.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11733, 286, 306570
Target Species Human, Mouse, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.