Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ankyrin 3 Antibody, PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305528PCP
Description
Ankyrin 3 Monoclonal antibody specifically detects Ankyrin 3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
Ankyrin 3 | |
Monoclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
ankyrin G, Ankyrin G119, ankyrin-3, ankyrin-G, Node Of Ranvier | |
The C-terminal 398 amino acids of human Ankyrin 3 expressed in and purified from E. coli. | |
0.1 mL | |
Cellular Signaling, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers | |
288 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
PerCP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction