Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ankyrin 3 Antibody (S106-20), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259310AF488
Description
Ankyrin 3 Monoclonal specifically detects Ankyrin 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Ankyrin 3 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ANK3 | |
Fusion protein 1000 C-terminal amino acids of human Ankyrin G encompassing all of Ankyrin G with the exception of Ankyrin repeats | |
100 μL | |
288 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
S106-20 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
ankyrin G, Ankyrin G119, ankyrin-3, ankyrin-G, Node Of Ranvier | |
Mouse | |
Protein G purified | |
Primary | |
Detects >200kDa. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction