Learn More
Ankyrin 3 Antibody (S106-20), DyLight 680, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259310FR
Catalog No. NBP259310FR
Only null left
Add to Cart
Description
Description
Ankyrin 3 Monoclonal specifically detects Ankyrin 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Ankyrin 3
Monoclonal
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
ANK3
Fusion protein 1000 C-terminal amino acids of human Ankyrin G encompassing all of Ankyrin G with the exception of Ankyrin repeats
100 μL
288
Human, Mouse, Rat
Purified
Western Blot, Immunocytochemistry, Immunofluorescence
S106-20
Western Blot, Immunocytochemistry/Immunofluorescence
ankyrin G, Ankyrin G119, ankyrin-3, ankyrin-G, Node Of Ranvier
Mouse
Protein G purified
Primary
Detects >200kDa.
Store at 4C in the dark.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.