Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ankyrin Brain Antibody (S105-13), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259374C
Description
Ankyrin Brain Monoclonal specifically detects Ankyrin Brain in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Ankyrin Brain | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ANK2 | |
Synthetic peptide amino acids 203-496 of human Ankyrin-B | |
100 μL | |
287 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S105-13 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Ank2, Ankyrin 2, Ankyrin 2 brain, Ankyrin B, Ankyrin brain, Ankyrin nonerythroid, Brain Ankyrin | |
Mouse | |
Protein G purified | |
Primary | |
Detects 200kDa. No cross reactivity against Ankyrin-G. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction